Extended Light Path Capillaries, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Extended light path capillaries that improve sensitivity 3- to 5-fold over standard capillaries.
- Use narrow 25 and 50 µm Int.Ø 'bubble' cell capillaries for highly conductive buffers without sacrificing sensitivity
With extended light path capillaries, the inner diameter is increased only at the detection window, offering the sensitivity of a wide inner diameter capillary and the low current generation of a narrow one. Resolution is not sacrificed when used with matching Agilent optical alignment interfaces.
Customers Who Bought This Also Bought