CrossLab RID Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
High-quality supplies for refractive index detectors, including tubing kits and capillaries.
- Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
- Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent provides high-quality replacement parts and supplies for refractive index detectors (RID). The robust and secure RID tubing kit includes recycle and purge valves in a range sizes. The interface tubing kit consists of replacement ferrules, nuts, and PTFE tubing required for accurate and efficient RID performance. A variety of interface and restriction capillaries are also available.
All RID replacement parts and components are manufactured to Agilent's quality standards to ensure they deliver excellent performance every time.
Customers Who Bought This Also Bought