CrossLab ISIS 3 (Integrated Sample Introduction System) Supplies for ICP-MS (7800/7900/8900), Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Supplies and sample loops for Agilent ISIS 3 switching valve used with 7800, 7850, 7900 and 8900 ICP-MS.
- ISIS 3 provides simpler operation, higher sample throughput and improved long term stability
- Complete kit of standard consumable parts for ISIS high sample throughput discrete sampling are provided
- A selection of sample loop sizes to suit your application needs are available and customizable to give you complete flexibiliy and optimal sample throughput
- Rotor seals, nuts and ferrules for valves can be provided
The ISIS 3 increases productivity when used with the Agilent 7800, 7850, 7900 and 8900 ICP-MS. ISIS 3 also allows high matrix samples to be run for extended periods of time. The ISIS 3 enables reduced sample uptake and rinse times with discrete sampling (DS), and delivers unmatched productivity for high sample matrix analysis in high throughput laboratories.
Agilent's ISIS 3 supplies ensure your laboratory productivity. Available switching valve parts include tubing, ferrules, and fittings for high throughput discrete sampling with ICP-MS.
Customers Who Bought This Also Bought