CrossLab Filters for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Water strainer, air inlet and coolant filter for Agilent ICP-MS instrument and recirculating chiller.
- Replacement water strainer for the ICP-MS cooling fluid supply removes large particulates to prevent problems with clogging
- Air inlet filter captures laboratory dust and debris to protect and prolong the instrument lifetime
- Check, clean and replace ICP-MS and recirculating chiller filters regularly to maintain performance
Replacement water strainer and air inlet filter for the ICP-MS instrument. These filters remove particulates and purify the cooling fluid to prevent problems with clogging and inadequate cooling due to restricted flows. The filters are designed to optimize ICP-MS system performance, providing a cost-effective safety measure to capture laboratory dust and debris. The water strainer and air filter protect and prolong the instrument lifetime.
Customers Who Bought This Also Bought