CrossLab Demountable Torch Supplies for 5100/5110 and 5800/5900 ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Injectors, outer tube sets, and other supplies for the fully demountable torch.
- Fully demountable torch provides the flexibility of having a variety of injector sizes and materials to handle difficult and diverse application demands
- Ease of maintenence with a tool free removable injector, all with no optimization required
- New organics outer tube set is made of high purity quartz providing longer lifetime when using organic solvents
- The vertical torch orientation provides the ultimate configuration for determining tough samples
- Handles even the toughest samples and achieves unmatched long term stability
- The plug-and-play torch eliminates the torch alignment process
- Ensures fast start-up and reproducible results with each operator
The new fully demountable torch for the 5000 Series ICP-OES can be used with a variety of injector sizes and materials to analyze any sample matrix. Outer tube sets are available in axial and radial configurations with an organics option available. The organics outer tube set extends the service life of the tube set when used with organic applications. The fully demountable torch features a removable injector to simplify torch maintenance and allows quick changeover for diverse sample matrices. The complexity of changing the injector when switching applications or cleaning has never been easier.
Customers Who Bought This Also Bought