Autosampler Sample Vials, Beakers and Adapters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Sample vials, beakers and adapters for use with Agilent PSD 100/120 Programmable sample dispenser.
- Sample vials are of heavy-duty construction to prevent cracking or tipping
- The conical design of the vials allows for use of nearly the entire sample
- Sample vials are molded from translucent, LDPE and are recommended for use with most aqueous solutions
- There is a choice of vials available for either the standard 50 position carousel (2 ml capacity) or the optional 130 sample high capacity carousel (1.1 ml capacity)
- Larger volume reagent containers are suitable for holding bulk quantities of blank solution for dilution and standard preparation, bulk standard for standard preparation, other reagent solutions and chemical or matrix modifiers
Agilent provides a range of supplies for the PSD 100/120 Programmable sample dispenser used with Agilent GFAAS systems, including sample vials, beakers, adapters, dispensing capillary assemblies, dispensing syringe and replacement plungers.
Adapters are also available to enable smaller sample vials to be located in the larger carousel positions when working with smaller volumes of chemical modifiers or other reagents.
Customers Who Bought This Also Bought