About this item
Safety and preventative measure assure the highest purity gas/air to the ICP-OES instrumentation.
- Delivers clean gases for accurate analyses.
- Both inlet and outlet connectors are equipped with stainless steel frits to prevent particulates from entering the gas stream.
- Air inlet filter captures laboratory dust and debris to protect and prolong the instrument.
- Easily replaced.
Agilent high-performance gas purifiers are designed to optimize ICP-OES system performance. Cartridges remove impurity gases, moisture, and hydrocarbons from ICP-OES gas supplies. The ICP-OES air inlet filter protects the instrument from debris and particles in the lab environment from entering the system. Replacement reminder tags ensure sensitivity and results aren’t impacted for optimal sensitivity and results. Agilent gas purifying cartridges for ICP-OES are a cost-effective safety and preventative measure to assure the supply of the highest purity gas to your ICP-OES instrumentation. Air inlet filter captures laboratory dust and debris to protect and prolong the instrument.