CrossLab Pump Supplies for Waters LC Systems, Agilent Technologies
Catalog #: CAAG8005-0513
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Agilent check valve cartridge, for Waters 1515/1525/515/510/2695D/600E/610/LC Module 1 pumps, similar to Waters 700000254
- Cat. no.:CAAG8005-0513
Specifications
About this item
Supplies for maintaining selected Waters HPLC systems, including common pump replacement parts.
- Agilent CrossLab parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent
Agilent pump supplies for selected Waters HPLC systems contain the most important components of a pump: plungers, seals, and check valves. Agilent HPLC supplies are manufactured and rigorously tested to ensure performance with your analytical instrument.
Regular pump maintenance helps lower operating costs and ensure precise and consistent results. By following a regular maintenance routine, you can count on maximum uptime, steady and accurate solvent flow, and pressure stability throughout the lifetime of the pump.
Customers Who Bought This Also Bought