CrossLab Graphite Furnace AAS Graphite Electrodes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Spare graphite furnace electrodes for optimum performance with maximum tube life.
- Keeping spare electrodes on-hand helps you maximize uptime
- Good electrical contact ensures optimum performance and maximizes tube life
- High purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise ratio
Agilent graphite furnace AAS electrodes provide good electrical contact, ensuring you achieve optimum performance with maximum tube life.
Customers Who Bought This Also Bought