Five-Slide Mailer, Caplugs
Catalog #: 76502-820
Supplier: Caplugs
Specifications
- Color:Natural
- Description:Polypropylene slide mailer for 5 slides
- Cat. no.:76502-820
Specifications
About this item
The Five-Slide Mailer features an integral recessed lid design which limits slide movement during transit to prevent accidental breakage.
- Rounded corners allow easier handling
- Eyelet is provided for optional security ties and/or identification tags
- Mailers can also function as miniature Coplin jars
Lightweight design (less than ¹/₃ oz.) helps to minimize transport costs.
Customers Who Bought This Also Bought