Specifications
- Description:Aerosol evacuation system for CytoFLEX SRT
- Cat. no.:CA76521-330
Specifications
About this item
The Aerosol Evacuation System removes aerosols and micro droplets, which may be generated during the course of normal operation or sort failure conditions, from the sort chamber without disturbing sorting.
- Ultra-quiet motor (40 dB Min and 69.5 dB Max) and a high-suction, high flow rate centrifugal action pump
- Micro droplets and particulates greater than 0.12 μm are removed under vacuum and trapped in an Ultra-Low Penetration Air (ULPA) filter
- Air flow rate is fully user adjustable, providing clearance of the sort chamber at rates of 10 to 40 complete air exchanges per minute
Aerosol Evacuation System was designed with operator safety in mind. It provides complete exchange of the air inside the sort chamber, filtering micro-droplets and airborne particulates with a replaceable ULPA filter. The system has an in-line evacuation motor that operates in two vacuum modes. These dual motor speeds provide complete evacuation of the sort chamber at the rate of 5-7 complete air exchanges per minute in low-speed mode, and 13-17 complete air exchanges per minute in high-speed mode.
The Aerosol Evacuation System is operated in low-speed mode during the sort process. This airflow rate in the sort chamber does not interfere with sorting, drop delay determination or droplet deposition. High-speed mode is used after completing the sort, to quickly exchange air inside the sort chamber prior to opening the aerosol containment door.