About this item
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
- 6" 2-ply edges resist punctures
- Reduces labor costs
- Simplifies the conformance to wrapping SOP's
- Steam sterilization indicator
- Autoclavable at 121 to 125 °C
- Hydrophobic
- Lint-free
- Manufactured and packaged in a controlled cleanroom environment
Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilization. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.
After sterilization and after the outer Tyvek® bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed. Remaining in place over the bowl during setup, the cover minimizes microbial or particulate contamination risks.