Spouts for Jerricans, S.C.A.T
Supplier: SCAT AMERICAS, INC.
Certificates
About this item
For simple and safe pouring of fluids from containers.
- Plastic
- Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S 55, S 60/61, and S 40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
Customers Who Bought This Also Bought