Orum International In-Rest Aspirating Head/Plate Holder, Hardy Diagnostics
Catalog #: CA76509-266
Supplier: HARDY DIAGNOSTICS CA
Specifications
- Dimensions:90x120x105 mm
- Description:In-Rest aspirating head/plate holder
- Cat. no.:CA76509-266
Specifications
About this item
To facilitate operations and avoid errors and risks of contamination during the sampling phase, the TRIO.BAS™ IN-REST is particularly useful as temporary support for culture media plates and/or lids, and instrument sampling heads.
- Constructed from AISI 316 stainless steel
- Wedge shape inclined at 150°
- Autoclavable
- Two-sided support
- Usable for Petri dishes, contact plates, and aspirating heads
- A single hole on one side, and two holes on the other side, facilitate identification of plates and heads
Operators, and their activities, represent the greatest risk of contamination inside cleanrooms. Operators must work under stringent conditions, as they are required to wear special overalls, gloves, facemasks, and eye masks. For these reasons, it is advisable to create the best possible working conditions. Air sampling with microbiological samplers is a complex and delicate operation, whether using an instrument with a single aspirating head, or an instrument with more than one aspirating head.
Customers Who Bought This Also Bought