CrossLab Nitrogen Chemiluminescence Detector Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Supplies for 255 Nitrogen Chemiluminescence Detector (NCD). This nitrogen-specific detector is based on a chemiluminescent reaction so complex sample matrices can be analyzed.
- Complex sample matrices can be analyzed with little or no interference
The Agilent 255 Nitrogen Chemiluminescence Detector (NCD) is a nitrogen-specific detector that produces a linear and equimolar response to nitrogen compounds based on a chemiluminescent reaction of NO with ozone. Even complex sample matrices can be analyzed with little or no interference.
Customers Who Bought This Also Bought