CrossLab Cleaning and Maintenance Supplies for LC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Agilent supplies a range of cleaning and maintenance supplies for your LC/MS.
- Preventative maintenance kits for modules of the Agilent 1220, 1260, and 1290 Infinity LC systems, as well as extended preventative maintenance kits for 1200, 1100, and 1050 Series pumps
- Repair kits for flow cells
- Abrasive powder, cleaning wires, and cloth for routine maintenance;Ball, nut, and torx drivers available
- Performance maintenance kits for Edwards and MS40+ kits
Agilent LC/MS preventive maintenance kits contain supplies commonly recommended to mantain Agilent LC/MS systems. Agilent also provides a range of specialized cleaning supplies including cloths, swabs, and dielectric capillary cleaning powder, to help your ensure systems operate at peak performance.
Customers Who Bought This Also Bought