CrossLab RF Coils for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Radio Frequency (RF) work or Load Copper Coil for Agilent 7700, 7800, 7900, 8800 and 8900 ICP-MS.
- ICP-MS radio frequency (RF) work load coil is part of the shield torch system
- RF copper coil is water cooled and should be replaced if signs of copper corrosion are evident or there is any physical damage to the coil
Replacement Agilent 7700, 7800, 7900, 8800, 8900 ICP-MS radio frequency (RF) work load coil, which is part of the shield torch system. The RF work load coil works together with plasma torch, long-life shield plate and bonnet.
The quartz torch for the ICP-MS is located in the center of a cooled copper work coil, through which a high power, high frequency electric current is passed. This creates an intense magnetic field inducing collisions between free electrons and Ar atoms, producing ions and more electrons, and creating or sustaining a stable, high temperature plasma.
Customers Who Bought This Also Bought