Imapure® Platinum Cured Silicone Transparent Tubing, Foxx Life Sciences
Supplier: Foxx Life Sciences
Certificates
About this item
Imapure® Platinum cured silicone transparent tubing, with ultra smooth bore is FDA 21 CFR< UPS Class VI Complient.
- Manufactured using high quality platinum cured grade silicone
- Manufactured and Packaged in dust free area of ISO 9001 QMS, ISO 14001 and OHSAS 18001 certified facility
- Increased fluid transfer characteristics than peroxide silicone tubing
- Superior transparency than peroxide tubing
- Heavy metal free
Designed for peristaltic pump and fluid transfer in pharmaceutical and biotech applications. Imapure™ is translucent silicone tubing known for its ultra smooth bore to prevent any particle entrapment. It is especially designed for the requirements of pharmaceutical needs.
Certifications: Complies with FDA 21 CFR, USP Class VI and ISO 10993.
Specifications
- Catalog No:
- 76517-518
- 76517-520
- 76517-436
- 76517-438
- 76517-504
- 76517-506
- 76517-440
- 76517-442
- 76517-508
- 76517-510
- 76517-512
- 76517-514
- 76517-516
- 76517-424
- 76517-426
- 76517-428
- 76517-430
- 76517-432
- 76517-434
- 76517-458
- 76517-460
- 76517-462
- 76517-444
- 76517-446
- 76517-448
- 76517-450
- 76517-452
- 76517-454
- 76517-456
- 76517-484
- 76517-320
- 76517-322
- 76517-304
- 76517-480
- 76517-482
- 76517-306
- 76517-308
- 76517-310
- 76517-312
- 76517-314
- 76517-316
- 76517-318
- 76517-464
- 76517-466
- 76517-468
- 76517-470
- 76517-472
- 76517-474
- 76517-476
- 76517-478
- 76517-264
- 76517-342
- 76517-260
- 76517-326
- 76517-502
- 76517-328
- 76517-262
- 76517-330
- 76517-332
- 76517-334
- 76517-336
- 76517-338
- 76517-340
- 76517-486
- 76517-488
- 76517-490
- 76517-492
- 76517-494
- 76517-496
- 76517-254
- 76517-498
- 76517-256
- 76517-258
- 76517-500
- 76517-324
- 76517-284
- 76517-286
- 76517-348
- 76517-282
- 76517-350
- 76517-352
- 76517-354
- 76517-356
- 76517-358
- 76517-360
- 76517-362
- 76517-266
- 76517-268
- 76517-270
- 76517-272
- 76517-274
- 76517-276
- 76517-344
- 76517-278
- 76517-280
- 76517-346
- 76517-370
- 76517-372
- 76517-374
- 76517-376
- 76517-378
- 76517-380
- 76517-382
- 76517-288
- 76517-290
- 76517-292
- 76517-294
- 76517-296
- 76517-364
- 76517-298
- 76517-366
- 76517-300
- 76517-302
- 76517-368
- 76517-392
- 76517-394
- 76517-396
- 76517-398
- 76517-400
- 76517-402
- 76517-384
- 76517-386
- 76517-388
- 76517-390
- 76517-414
- 76517-416
- 76517-418
- 76517-420
- 76517-422
- 76517-404
- 76517-406
- 76517-408
- 76517-410
- 76517-412
- Sterilisation:
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation
- Autoclaving; Ethylene oxide; Gamma radiation