About this item
Wide pore superficially porous particles with C3, C8 and C18 ligands available.
- UHPLC separations of biomolecules with superficially porous particles
- 300Å pore provide high efficiency and recovery with proteins (up to 1000 kDa) and monoclonal antibodies
- Achieve long lifetime at low pH with Poroshell 300SB at high pH with 300Extend-C18
- Optimize recovery and selectivity with four different bonded phases – 300SB-C18, 300SB-C8, 300SB-C3, and 300Extend-C18
Agilent Poroshell columns are ideal for fast separations of proteins and peptides because the unique particle allows for fast flow rates to be used while maintaining sharp, efficient peaks. Peptides and proteins are typically separated slowly to reduce the potential peak broadening of these slow diffusing analytes. But Poroshell columns use a unique particle made with a layer of porous silica on a solid core of silica. This reduces the diffusion distance for proteins making practical rapid HPLC separations of peptides and proteins up to 500–1,000 kDA. Poroshell 300SB C18, C8, C3, and 300Extend columns provide excellent stability and selectivity choices with TFA and formic acid mobile faces. Poroshell 300Extend C18 columns can be used at varying pH levels for unique separations and additional selectivity choices.
Specifications
- Catalog No:
- CA821075-924
- CA821075-918
- Max. pressure:
- 400 bar
- 400 bar
- pH range:
- 1-8
- 1-8
- Temperature limit:
- 90 °C
- 90 °C
- Wetted materials:
- Methanol/Water