J&W Ultra-1 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A nonpolar column with tighter QC specifications that delivers excellent accuracy in a range of chromatographic applications.
- 100% Dimethylpolysiloxane
- Nonpolar
- Equivalent to HP-1, with tighter specifications for retention index and capacity factors
- Bonded, crosslinked, and solvent rinsable
- Intended for use with the Sadtler Retention Index Library
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W Ultra 1 is a 100% dimethylpolysiloxane, nonpolar column that resembles HP-1 but has tighter specifications for retention index and capacity factors. The column has an operating temperature range of –60 - 325/350 °C and is intended for use with the Sadtler Retention Index Library.
These precision-engineered columns are bonded, crosslinked, and solvent rinsable, and are tested to some of the strictest industry QC standards, ensuring confidence in the accuracy and integrity of analytical results.
Specifications
- Catalog No:
- CAAG19091A-112
- CAAG19091A-115
- CAAG19091A-005
- CA19091A-101
- CAAG19091A-012
- CAAG19091A-102
- CAAG19091A-105
- CAAG19091A102E
- CAAG19091A-002
- CAAG19091A101I
- CAAG19091A-008
- CA19091A-108
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase