J&W CP-Molsieve 5Å Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Delivers highly efficient and accurate separation of argon and oxygen at ambient temperature.
- Separates argon and oxygen at ambient temperature to reduce costs
- High efficiency for increased productivity
- Symmetrical peaks for accurate results
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W CP-Molsieve 5 Å is a molecular-sieve-coated PLOT capillary column specially suited for high-resolution separation of permanent gases. Analysis times are reduced by up to 75% compared to packed columns. Baseline separation of Ar/O₂ is achieved at ambient temperature. The column's thin-layer dimensions produce fast elution of CO with symmetrical peaks.
Customers Who Bought This Also Bought