J&W DB-ProSteel Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Virtually unbreakable metal columns that provide excellent peak shapes. Available in a wide variety of stationary phases.
- Excellent peak shapes
- Virtually unbreakable
- Available in a wide variety of stationary phases
- Bonded and crosslinked
- Ideal for high-temperature analysis and process applications
- Superior replacement for MXT/Silco-steel columns
Agilent J&W DB-ProSteel metal columns are engineered to combine the robustness of stainless steel with advanced surface deactivation for excellent peak shape. The columns can be custom-wound on request for small GC ovens. DB-ProSteel has the same outer diameter as standard megabore columns of 0.53 mm int.Ø so no special ferrules are required for proper installation.
Specifications
- Catalog No:
- CAAG145-1334
- CAAG145-2814
- CAAG145-7032
- CA145-1011
- Polarity:
- Mid-polarity stationary phase
- Low-polarity stationary phase
- High-polarity stationary phase
- Low-polarity stationary phase
- USP code:
- G43
- G14, G15, G16, G20, G39, G47
- G1, G2, G9, G38