PLRP-S for Biomolecules, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Agilent now offers PLRP-S for Biomolecules HPLC Columns. Please visit this PLRP-S for Biomolecules HPLC Columns page to learn more about PLRP-S for Biomolecules HPLC Columns.
- Contain durable and resilient polymer particles that deliver reproducible results over longer lifetimes
- Thermally and chemically stable
- Comply with USP L21 designation
- Used in bioscience, chemical, clinical research, energy, environmental, food and agriculture, material science and pharmaceutical industries
- Pore sizes (100Å-4000Å) for separations of small molecules to large complexes and polynucleotides
PLRP-S for Biomolecules products are available in a range of pore sizes and particle sizes, all with identical chemistry and fundamental adsorptive characteristics. The particles are inherently hydrophobic so there is no bonded phase, alkyl ligand required for reversed-phase separations.
This gives a highly reproducible material that is free from silanols and heavy metal ions. Within the extensive product range are columns suitable for analytical separations, and preparative purifications. In addition, process columns can be packed with the bulk media.
Specifications
- Catalog No:
- CAAGPL19123801
- CAAGPL19121803
- CAAGPL15121300
- CAAGPL19123300
- CAAGPL15121802
- CAAGPL19121501
- CAAGPL12123201
- CAAGPL19121502
- CAGPL19122502
- CAAGPL19123501
- CAAGPL15125801
- CAAGPL15121803
- CAAGPL15123803
- CAAGPL15121501
- CAAGPL15125800
- CAAGPL19121500
- CAAGPL15123801
- CAAGPL13121802
- CAAGPL15123300
- CAAGPL15123800
- CAAGPL15123301
- CAAGPL15121500
- CAAGPL15121301
- Max. pressure:
- 207 bar
- 207 bar
- 275 bar
- 275 bar
- 207 bar
- 207 bar
- 103 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 207 bar
- 275 bar
- 207 bar
- 275 bar
- 207 bar
- 275 bar
- pH range:
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- Temperature limit:
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 80 °C
- 80 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- Wetted materials:
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water
- Acetonitrile/Water