J&W CP-Sil 88 for FAME Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Guarantees analysis of FAME cis/trans isomers in the C6 to C26 range.
- Optimized for analysis of FAME cis/trans isomers
- High-polarity stationary phase increases efficiency and productivity
- Use for FAME separations in the C6 to C26 range
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W CP-Sil 88 for FAME analysis delivers resolution of cis/trans FAME isomers for complete confidence.
Specifications
- Catalog No:
- CAAGCP7488
- CAAGCP7489I5
- CACP7489
- CAAGCP7487
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- USP code:
- G5
- G5
- G5