About this item
Superficially porous particle columns with high pH stability for oligonucleotide analysis by IP-RPC.
- Agilent Bioconsumable standards are of the highest quality and are designed, developed, and manufactured to guarantee at least one year of shelf life.
To successfully separate trityl-off, deprotected oligonucleotides, you need columns that have high resolving power and are robust enough to withstand challenging conditions.
If your columns don’t provide enough resolution, you can’t be sure your results are accurate. If your columns are not robust, you will have to replace them frequently, which will disrupt your workflow and increase your costs.
Agilent AdvanceBio oligonucleotide columns feature high efficiency, 2.7 μm superficially porous Poroshell particles. Using technology exclusive to Agilent, we chemically modify the particles to make them exceedingly resistant to high pH mobile phases. We also bond them with an end-capped C18 phase that delivers excellent selectivity for oligonucleotides. What’s more, we test every batch of AdvanceBio oligonucleotide media with a resolution standard to ensure consistent performance.
The family of AdvanceBio products are designed to deliver consistent, exceptional performance for the complete characterization of proteins, antibodies, conjugates, new biological entities and biopharmaceuticals.
Specifications
- Catalog No:
- CAAG821725-921
- CAAG653750-702
- Max. pressure:
- 600 bar
- 600 bar
- pH range:
- 3-11
- 3-11
- Temperature limit:
- 65 °C
- 65 °C
- Wetted materials:
- Acetonitrile/Water
- Acetonitrile/Water