CrossLab Direct Connect Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Direct Connect liners provide sensitivity,maximum recovery, and minimal decomposition of active compounds for methods requiring splitless injection such as EPA 8270.
- Press fit connection to the column
- No sample exposure to inlet
- Included in 8270 EPA application kit
- Optimized hole size and placement
Agilent's Direct Connect liners provide maximum recovery and minimal decomposition of active compounds for methods requiring splitless injection, such as EPA 8270. They are best for relatively clean samples containing active analytes, such as water extracts. The liners directly connect with the column, similar to press-fit connectors, to aid complete transfer of sample onto the column, eliminating the problem of inlet discrimination and further increasing sensitivity. Direct connect liners are not designed for the purposes of high volume injections.
Customers Who Bought This Also Bought