ContactUsLinkComponent
Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Supplier:  VWR International
   Sustainable Options Available
Certificates
Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Supplier:  VWR International
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 76514-184
  • 76514-180
  • 76514-182
  • Environmentally Preferable:
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Cat. No.:
  • 76514-184
  • 76514-180
  • 76514-182
  • Defrost:
  • Controlled auto defrost CAD (freezer); Cycle defrost (refrigerator)
  • Manual defrost (freezer); Cycle defrost (refrigerator)
  • Auto defrost (freezer); Cycle defrost (refrigerator)
  • Shelves/drawers:
  • 3 adjustable shelves (refrigerator) / 2 adjustable shelves (freezer)
  • 3 adjustable shelves (refrigerator) / 2 adjustable shelves (freezer)
  • 3 adjustable shelves (refrigerator) / 2 adjustable shelves (freezer)
  • Door type:
  • 1 solid swing door (refrigerator); 1 solid swing door (freezer)
  • 1 solid swing door (refrigerator); 1 solid swing door (freezer)
  • 1 solid swing door (refrigerator); 1 solid swing door (freezer)
  • Electrical:
  • 115 V, 60 Hz; 1.3 A, 1/6 HP refrigerator; 1 A, 1/8 HP freezer
  • 115 V, 60 Hz; 1.3 A, 1/6 HP refrigerator; 1 A, 1/8 HP freezer
  • 115 V, 60 Hz; 1.3 A, 1/6 HP refrigerator; 1 A, 1/8 HP freezer
  • Weight:
  • 97 kg (215 lbs.)
  • 97 kg (215 lbs.)
  • 97 kg (215 lbs.)