CleanMax® Clean Manufactured Non Sterile Sleeves, Lakeland Industries
Catalog #: 76492-452
Supplier: Lakeland
Specifications
- Length:18"
- Color:White
- Description:Clean manufactured sleeves with tunneled elastic ends
- Cat. no.:76492-452
Specifications
About this item
Clean manufactured nonsterile sleeves with bound seams. Clean manufactured in a silicone-free environment.
- Fluid repellent; tested in accordance with ASTM D6978 for Chemotherapy Permeation
- Open fold for easier donning
- Resistant to blood, body fluid and viral penetration
- Resistant to blood borne pathogens
- IEST-RP-CC003 Category I Particle Cleanliness
- Latex-free and silicone-free
CleanMax® cleanroom garments for controlled environments provide optimal comfort, quality, and protection. Ideally suited for use in ISO Class 4 - 8 cleanrooms and all controlled environments. CleanMax® garments are easy to don, comfortable to wear, and are backed by Lakeland's 30+ years as a manufacturer of disposable protective apparel.
Packaging: Packaged in pairs in a protective outer bag for ante areas.
Customers Who Bought This Also Bought