extraktLAB Chromatography Media, Cannabinoid Remidiation, United Science
Catalog #: CA76501-280
Supplier: United Science
CAS Number:
Specifications
- Description:Chromatography media, 30 - 60 µm, 100 Å, 1 kg
- Cat. no.:CA76501-280
Specifications
About this item
Silica bonded phase used for THC-CBD separations, purification, and remediation.
Customers Who Bought This Also Bought