Specifications
- Description:Digital display ESD tester and footplate
- Cat. no.:76490-158
Specifications
About this item
This OHM Metrics™ unit is a comprehensive, accurate ESD ground device tester that fully meets daily ESD checking needs.
- Tests wrist straps and footwear
- Proactive 'near-fail' alert indicates if a ground device is close to fail levels
- Eliminate failures in the field
- Preset with broad selectable test range: 100 KΩ to 1 GΩ
An easy, accurate, and time saving one-touch digital wrist strap and footwear tester. Simultaneously and independently test both feet and a wrist strap while filtering out human body noise. The results are visible via a digital screen. A unique 'near-fail' indicator alerts the user that the ESD grounding device under test (wrist strap or heel grounder) is nearing failure levels and should be changed soon, if not right away. This helps prevents a worn device 'passing' a test and failing in the field. With the digital readout, the user will know excally how worn a device is. This tester fully supports the 'Compliance Verification Plan' requirements as stated in ANSI ESD S20.20