Air Ionizing Gun, Transforming Technologies
Catalog #: 76489-744
Supplier: Transforming Technologies
Specifications
- Description:Ac ionizing gun
- Cat. no.:76489-744
Specifications
About this item
A lightweight but durable static removal gun.
- Point and shoot precision ionization
- Durable impact-resistant fiberglass reinforced case
- Quick disconnect air fittings
The ionizing gun provides superior control and targeted particulate cleanup for a wide variety of industrial applications. As a forceful neutralizer and blow-off gun, it easily removes stubborn particulate from surfaces. As a neutralizer, it gets rid of troublesome charges responsible for the attraction of contaminants to product surfaces.
Customers Who Bought This Also Bought