Static Field Meter, Transforming Technologies
Catalog #: 76489-642
Supplier: Transforming Technologies
Specifications
- Description:Static field meter
- Cat. no.:76489-642
Specifications
About this item
Perform static surveys and evaluate charge accumulation in electronic manufacturing and industrial operations such as converting, printing, packaging, and plastics processing.
- LED guide: pulsing-beam rangefinder to consistently obtain correct readings
- HOLD button captures transient readings
- Measuring range: + 0 ~ 20 kV
The Static Field Meter is used to perform static surveys and evaluate static charge accumulation. The compact design and four button operation make it easy to operate. It can measure static charges up to 20 kV. Simply hold the instrument so the LED guide beams form a non-pulsing circle indicating an exactly distance from the target.
Customers Who Bought This Also Bought