Wolfgang Warmbier Backrest Probe, Transforming Technologies
Catalog #: 76489-574
Supplier: Transforming Technologies
Specifications
- Description:Backrest probe
- Cat. no.:76489-574
Specifications
About this item
The Backrest probe is designed to evaluate ESD chair backrests according to ANSI ESD20.20.
- Ensures excellent contact during evaluations of ESD chairs
- Rugged metal probe
- 4 mm banana plug socket connection
For use with resistance meters.
Customers Who Bought This Also Bought