Wolfgang Warmbier Glove Probe, Transforming Technologies
Catalog #: 76489-570
Supplier: Transforming Technologies
Specifications
- Description:Glove probe
- Cat. no.:76489-570
Specifications
About this item
Glove probe is designed to evaluate ESD gloves according to ANSI ESD20.20.
- Ensures excellent contact during evaluations of ESD gloves
- Rugged metal probe is fitted with conductive contact points for resistance meter cable
- 4 mm banana plug socket connection
The probe’s weight ensures excellent contact on ESD glove measurements. The rugged clamp is fitted with conductive contact points for a resistance meter cable. The connection point is a 4 mm banana plug socket.
Customers Who Bought This Also Bought