Wolfgang Warmbier Charge Plate Monitor, Transforming Technologies
Catalog #: 76489-528
Supplier: Transforming Technologies
Specifications
- Description:Charge plate monitor
- Cat. no.:76489-528
Specifications
About this item
The Warmbier Charge Plate Monitor is a simple-to-use device to measure an air ionizer's performance.
- Test ionizer decay times and balance according to ANSI ESD2020
- Simple to use two button operation
- Compact and portable
Quickly and easily test ionizer decay times and offset voltage (balance) according to ANSI ESD2020 and IEC 61340-2-1. Powered by an internal rechargeable battery in a compact, easy to carry design. An optional high voltage head allows direct voltage measurement of up to 2 kV (sold separately).
Customers Who Bought This Also Bought