LiquidGuard™ 3 Pocket Lab Coats, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Certificates
About this item
LiquidGuard™ 3 Pocket Lab Coats are impervious yet breathable, making them comfortable to wear for extended periods.
- Manufactured from microporous breathable film laminated non-woven fabrics
- Knee length, 3 pockets, snap front closure, knit cuffs, and knit collar
- Lint-free, low particle shedding, rated category 1 per Helmke Drum Test
- Latex-free, TSE-free, BSE-free, and silicone-free
- Manufactured in a controlled environment
LiquidGuard™ 3 Pocket Lab Coats pass blood penetration test per ASTM F1670, pass ChemoDrug Permeation Test per ASTM F729-12, and pass permeation tests of various non-hazardous laboratory chemicals. Offering a high vapor/air transmission rate, they are breathable and comfortable to wear.
These lab coats are ideally suited for use in hospital, industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Certifications: Manufacturing Facility is ISO 9001 and ISO 13485 Certified and FDA Registered and Inspected.
Packaging: Lab coats are packaged 10 per pack, 50 per case.
Specifications
- Catalog No:
- 76488-490
- 76488-480
- 76488-478
- 76488-484
- 76488-482
- 76488-488
- 76488-486
- Bacterial filtration efficiency:
- 99%
- 99%
- 99%
- 99%
- 99%
- 99%
- 99%
- Particle shedding (Helmke Drum):
- Category I
- Category I
- Category I
- Category I
- Category I
- Category I
- Category I
- Water vapor transmission rate:
- 977 g/m²/24 h
- 977 g/m²/24 h
- 977 g/m²/24 h
- 977 g/m²/24 h
- 977 g/m²/24 h
- 977 g/m²/24 h
- 977 g/m²/24 h
- Weight:
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
- 56 g/m²
Specifications
Customers Who Bought This Also Bought