N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) silylation reagent, Pierce™
Catalog #: 10066-350
Supplier: Thermo Scientific
CAS Number: 77377-52-7
Specifications
- Size:5 mL
- Cat. no.:10066-350
- Supplier No.:TS-48920
Specifications
About this item
Synonyms: N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide
:Storage temperature: Ambient
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 241.33 g/mol
UN:1993
:3
:II
:GHS02
:GHS07
Formula:C₉H₁₈F₃NOSi
MW:241.33 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00009671
CAS Number:77377-52-7
Customers Who Bought This Also Bought