N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide with 1% tert-butylchlorodimethylsilane silylation reagent, Pierce™
Catalog #: 10066-352
Supplier: Thermo Scientific
CAS Number: 77377-52-7
Specifications
- Packaging:Ampoule
- Size:10 x 1 ml
- Cat. no.:10066-352
- Supplier No.:TS-48927
Specifications
About this item
Synonyms: N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide,Silylating mixture VI
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 241,33 g/mol
UN:1993
:3
:III
:GHS02
:GHS07
Formula:C₉H₁₈F₃NOSi
MW:241,33 g/mol
MDL Number:MFCD00009671
CAS Number:77377-52-7
Customers Who Bought This Also Bought