Urea
Catalog #: CA95017-130L
Supplier: Cytiva
CAS Number: 57-13-6
Specifications
- Size:500 g
- Cat. no.:CA95017-130L
- Supplier No.:17-1319-01
Specifications
Specification Test Results
- Urea:
Specification Test Results
About this item
Synonyms: Carbonyl diamide,Carbamide,Diaminomethanone
:Storage temperature: Ambient
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.34 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 197 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 132.7 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.06 g/mol
Merck Index:12,10005
Formula:NH₂CONH₂
MW:60.06 g/mol
EINECS:200-315-5
:197 °C (1013 hPa)
Storage Temperature:Ambient
MDL Number:MFCD00008022
CAS Number:57-13-6
REACH:01-2119463277-33
Customers Who Bought This Also Bought