Specifications
- Description:2-AB Man5/M5 N-glycan standard
- Size:100 pmol
- Cat. no.:CAGKSB-103
Specifications
About this item
2-AB labeled N-glycan standards and libraries provide high confidence in identification of samples. N-glycan (oligomannose 5) labeled with 2-aminobenzamide (2-AB).
- Commonly used as qualitative standards for LC/FLD or LC/MS
Agilent 2-AB (formerly ProZyme) labeled N-glycan standards and libraries are commonly used as qualitative standards for LC/FLD and LC/MS. These standards come pre-labeled with 2-AB, ready to reconstitute and inject.
Monoisotopic Mass: MWGlycan + 120.06875 = 1354.5021
High Mannose-type Native N-Glycan.