Cuvette Standard Q Optical Glass, Mettler-Toledo
Supplier: Mettler Toledo
Certificates
About this item
The Standard Q macro cuvette is made of fused quartz glass and comes in a matching pair. For measurements in the ultraviolet and visible range with a wavelength between 200 nm – 2500 nm.
- Reliable, Reusable Glass Cuvettes
Accuracy by design
To ensure high accuracy and reliability of measurements, standard cuvettes come in matching pairs and are manufactured within tolerances of ± 0.02 mm.
Clear view
Standard line cuvettes provide good optical transparency > 80% and chemical stability.
Robust and resistant
Standard Q cuvettes are seamlessly produced in a fire-fusing process which makes them robust and resistant even against the most corrosive chemicals.
Customers Who Bought This Also Bought