Vacuum Hydro Aspirator, Mortech®
Catalog # 76498-430
Supplier: Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Vacuum hydro aspirator
- Environmentally Preferable:
- Cat. no.:76498-430
Specifications
About this item
Deluxe hydro-aspirator with built-in vacuum breaker and control lever to be used to reverse the flow of water so as to direct it through the hose connections for either drawing fluid from the body cavity or rinsing a desired location.
- Fits most standard faucet threads
- Fabricated from chrome-plated brass
Unit can be purchased individually, or part of a whole water control unit.