Specifications
- Description:Stainless steel dissecting tray
- Cat. no.:76498-300
- L×W×D:25×18×1"
Specifications
About this item
A multi-purpose unit, this durable tray can be used to confine spills or contamination to a restricted area.
- All stainless steel
- Dissecting tray with sloped angle end for draining
All of Mopec’s standard dissecting boards will fit into this 1" deep tray. The sloped end overhangs any sink, allowing all fluids to drain directly into the sink.