Economy Platform Scale, Mortech®
Supplier: Mortech Manufacturing
Certificates
About this item
Economy platform scales are available in 2000 or 5000 g weighing capacities.
- 6" (15.2 cm) diameter platform
- Gram/lbs./oz. weighing modes, (1 g/0.05 oz. readability)
- Easily portable, tare function
- Battery or AC power options
- One year warranty
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.