LiquidGuard™ Sterile Sure-Clean Sleeve Covers, Apex Aseptic Products
Catalog #: 76488-320
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Specifications
- Clothing Size:21"
- Sterile:+
- Color:White
- Cat. no.:76488-320
Specifications
About this item
Sterile LiquidGuard™ sleeve covers are suitable for cleanroom use.
- LiquidGuard™ microporous fabric construction
- Impervious yet breathable
- Easy breathability, comfortable to wear
- Low particle shedding, rated category 1 per Helmke Drum Test
- Latex-free, TSE-free, BSE-free
- Manufactured in a controlled environment
- Gamma irradiated validated sterile to SAL 10⁻⁶
- Suitable to use in ISO Class 5 or higher cleanrooms
Sleeves feature attached thumb loops to help keep them in place. Tunneled sewing greatly reduces particle shedding. A special folding system helps to ensure proper aseptic donning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected. Pass Blood Penetration Test per ASTM F1670.
Delivery: Each package includes a gamma irradiation indicator dot, and each shipping case is affixed with a Certificate of Conformance and Certificate of Sterility.
Packaging: Sleeves are packaged 2 pieces per package, triple bagged, 200 pieces per case.
Customers Who Bought This Also Bought