Sterile Dupont® Tyvek™ Sleeve Covers, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Certificates
About this item
Sterile sleeve covers made with Dupont® Tyvek™ fabrics and are suitable for cleanroom use.
- Dupont® Tyvek™ fabrics
- Fluid resistant and breathable
- Low particle shedding, rated category 1 per Helmke Drum Test
- Latex-free, TSE-free, BSE-free
- Manufactured in a controlled environment
- Gamma irradiated validated sterile to SAL 10⁻⁶
- Suitable to use in ISO Class 5 or higher cleanrooms
Sleeves feature attached thumb loops to help keep them in place. Tunneled sewing greatly reduces particle shedding. A special folding system helps to ensure proper aseptic donning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected.
Delivery: Each package includes a gamma irradiation indicator dot, and each shipping case is affixed with a Certificate of Conformance and Certificate of Sterility.
Packaging: Sleeves are packaged 2 pieces per package, triple bagged.
Customers Who Bought This Also Bought