LiquidGuard™ Sterile Cleanroom Under-Chin Crown Hoods, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Certificates
About this item
Sterile LiquidGuard™ Under-chin crown hood for cleanroom use feature a crown top design, with elastic around the face to minimize skin exposure.
- LiquidGuard™ microporous material construction
- Impervious yet breathable
- Low particle shedding, rated category 1 per Helmke Drum Test
- Latex-free, TSE-free, BSE-free
- Manufactured in a controlled environment
- Gamma irradiated validated sterile to SAL 10⁻⁶
- Suitable to use in ISO Class 5 or higher Cleanrooms
These hoods feature tunneled elastic sewing to help reduce particle shedding. Elastic at the back of the head allows for a proper fit. A specially designed folding system ensures proper aseptic donning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected. Pass Blood Penetration Test per ASTM F1670.
Delivery: Each package includes a gamma irradiation indicator dot, and each shipping case is affixed with a Certificate of Conformance and Certificate of Sterility.
Packaging: Hoods are packaged 1 piece per package, triple bagged, 100 pieces per case.
Customers Who Bought This Also Bought