LiquidGuard™ Sterile Cleanroom Gowns, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Certificates
About this item
Sterile LiquidGuard™ cleanroom gowns are knee length, and feature a tie at back for secure closure.
- LiquidGuard™ microporous fabric construction
- Low particle shedding, rated category 1 per Helmke Drum Test
- Gamma irradiated validated sterile to SAL 10⁻⁶
- Suitable to use in ISO Class 5 or higher Cleanrooms
- Latex-free, TSE-free, BSE-free
Manufactured in a controlled environment, each garment is individually packaged with a gamma irradiation indicator dot.
These gowns feature a tie at the back around both the neck and waist, as well as elastic cuffs and thumb loops. The inside out folding system helps to achieve proper aseptic gowning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected. Pass Blood Penetration Test per ASTM F1670. Pass ChemoDrug Permeation Test per ASTM F739-12.
Delivery: Each case includes a Certificate of Conformance and Certificate of Sterility.
Packaging: Gowns are packaged 1 piece per package, triple bagged, 50 pieces per case.
Customers Who Bought This Also Bought