About this item
Sterile CoolKlean™ cleanroom coveralls feature a zipper front closure, without hood and booties.
- Made with high strength SMS fabrics
- High bacteria filtration rate: >92%
- High particle filtration rate: >93%
- Highly fluid resistant; Hydrohead >600 mm
- Easy breathability and comfortable to wear
- Zipper front, no hood, no booties
- Highly anti-static treated and latex-free
- Low particle shedding, rated category 1 per Helmke Drum Test
- Gamma irradiated, validated sterile to SAL 10⁻⁶
- Suitable to use in ISO Class 5 or higher cleanrooms
Inside seams are bonded with Teal color fabrics to indicate areas safe to touch inside the garment. Tunneled elastic cuffs with thumb loops help keep sleeves in place and prevent bare arms from exposure. Uniquely designed Clean-Don™ gowning system incorporates inside out folding and telescope leg method to ensure wearer’s hands are not contaminating the outside of the sterile garments and coverall’s legs are not touching the floor.
Each piece is individually packaged, triple bagged in plastic bags with easy tear notches, and labeled with item code, product name, lot number, production date, expiration date, and fixed with gamma irradiation indicator dot. Manufactured in a controlled environment.