Universal Columns, Caplugs
Supplier: Caplugs
Certificates
About this item
Universal columns have a multitude of applications in life sciences as well as in clinical applications.
- Polypropylene is chemically resistant to weak acid and alkali
- Total capacity of reservoir is 25 ml
Columns come complete with filter support in three levels of porosity: Fine (15 to 45 µm), medium (45 to 90 µm) or coarse (90 to 135 µm).
Customers Who Bought This Also Bought